Guard Cartridge Holder for HPLC Columns
Catalog # 80101/N
Supplier: Avantor
Specifications
- Popis:All-Guard cartridge holder for 2,1 to 4,6 mm ID columns
- Environmentally Preferable:Low Manufacturing Impact
- Certifikát ochrany životního prostředí:ISO 14001:2015 (530840 UM15)
- Bal.:1
Specifications
About this item
This guard cartridge holder promotes increased column lifetime.
- Improved hardware
- Higher pressure rating
- Low internal swept volume
The hardware of the Alltech All-Guard cartridge system has been improved and all guard cartridges and holders are now supplied in this new hardware, leading to more robust and higher efficiency separations. Chromatographic selectivity will be unaffected by this change.