Separating funnels, Squibb, HDPE stopper, PTFE stopcock
Katalogové číslo HACH52049
Dodavatel: Hach
Specifikace
- Kapacita:500 ml
- Bal.:1
Specifikace
O tomto produktu
Pear-shaped (Squibb) glass separatory funnel with PTFE stopcock and HDPE stopper.
- Clear glass body allows for easy detection of layers between immiscible liquids
- Highly chemical resistant and fully autoclavable
- HDPE stopper with closed bottom