Orders LinkContactUsLinkComponent
Forceps
Forceps
Straight, wide tips, 134,62 mm
Catalog # HACH2141100
Supplier:   Hach
Forceps
Catalog # HACH2141100
Supplier:  Hach
Forceps
Forceps
Straight, wide tips, 134,62 mm
Price
2 450,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Stainless steel.
  • Tips are rounded to prevent damage
  • Easy to use because of slip-proof plastic finger tips
  • Autoclavable
Specifications
  • Type:
    Straight, wide tips
  • Délka:
    134,62 mm