ContactUsLinkComponent
Cell dissociation reagent, Corning® Cellstripper®
Cell dissociation reagent, Corning® Cellstripper®
Catalog # 25-056-CI
Supplier:  Corning
CAS Number:  
undefined
Cell dissociation reagent, Corning® Cellstripper®
Catalog # 25-056-CI
Supplier:  Corning
CAS Number:  
Specifications
  • Bal.:
    6
  • Popis:
    Cellstripper®, liquid, 6×100 ml
  • Sekvence:
    45
Specifications
About this item
Cellstripper® is a non-enzymatic cell dissociation solution formulated with a proprietary mixture of chelators that gently dislodge adherent cells in tissue culture. It is an alternative to trypsin when used with either serum-free or serum-supplemented media. Because Cellstripper® is a gentler reagent than trypsin, cells can be exposed to Cellstripper for longer periods of time. Cellstripper® achieves superior results in comparison to enzymatic dissociation products.