Coliform swab
Catalog # 710-1479
Supplier: HYSERVE
Specifications
- Popis:Swabs for the detection of Coliforms
- Sekvence:04
- Bal.:25
Specifications
About this item
Easy to carry early warning test for contamination of surfaces with coliform bacteria. Based on a certified method, the Coliform Swab provides a simple method for the qualitative detection of these bacteria - with a clear colour change from purple to yellow in the presence of coliform bacteria.
- Simple colour change shows contamination, no colour change indicates the corresponding contamination
- Individually packed straightening test, which can be carried out without additional reagents
- Results can be read within 18 hours
- Determines the number of germs from 1 KbE/cm²
- Ideal for HACCP concepts
- Production certified according to ISO 9001
Escherichia coli and coliform bacteria are usually used as indicators for faecal impurities in water and other environmental samples. The presence of these bacteria usually leads to a lack of hygiene in a production section or to the contamination of water.
In order to confirm the absence of coliforms, the sample should be incubated for 24 hours so that even the smallest germ numbers can be detected.
Caution: *Výrobky na této stránce nespadají do oblasti působnosti směrnice EU 2019/904