Orders LinkContactUsLinkComponent
2X SDS-PAGE sample buffer without DTT or b-ME
2X SDS-PAGE sample buffer without DTT or b-ME
09, SDS-PAGE 2X sample buffer without DTT or b-ME (125 mM tris HCl, 20% glycerol, 4% SDS, 0,1% bromphenol blue, pH 6,8), 100 mL
Catalog # ROCKMB-018
2X SDS-PAGE sample buffer without DTT or b-ME
Catalog # ROCKMB-018
CAS Number:  
2X SDS-PAGE sample buffer without DTT or b-ME
09, SDS-PAGE 2X sample buffer without DTT or b-ME (125 mM tris HCl, 20% glycerol, 4% SDS, 0,1% bromphenol blue, pH 6,8), 100 mL
CAS Number:  
Price
4 350,00 Kč
Log in for availability
1 * 100 mL
Product Details & Documents
Sample buffer for protein electrophoresis.
This product is a concentrated stock solution and should be diluted appropriately with distilled, deionised water or equivalent to its final working concentration. 2X SDS-PAGE sample buffer consists of 0,125 M Tris, 4% (w/v) SDS, 20% (v/v) glycerol and 0,01% (w/v) bromophenol blue. The pH of this solution is 6,8. Meticulously prepared using ultra pure reagents dissolved in highly polished pharmaceutical grade deionised water.
This product was aseptically filtered through a Millipore 0,22 µm filter into clean, pre-sterilised containers. The product was tested on trypticase soy agar for 24 hours, 48 hours and 72 hours and was found to be negative for bacteria.
Caution: This product is for research use only and is not intended for therapeutic or diagnostic applications.
Documents
Specifications
Specifications
  • Bal.:
    100 mL
  • Popis:
    SDS-PAGE 2X sample buffer without DTT or b-ME (125 mM tris HCl, 20% glycerol, 4% SDS, 0,1% bromphenol blue, pH 6,8)
  • Sekvence:
    09