Insulin-Transferrin-Selenium (ITS) 100x Growth Supplement, Corning®
Catalog # MDTC25-800-CR
Supplier: Corning
CAS Number:
Specifications
- Typ balení:Plastová láhev
- Bal.:10 mL
- Popis:Insulin-Transferrin-Selenium (ITS) Growth Supplement, Sterile, 100X
- Sekvence:52
Specifications
About this item
Animal free formulation
Contains:
1000 mg/L insulin, human recombinant
550 mg/L transferrin, human recombinant
0.67 mg/L selenious acid
Caution: Storage 2°C to 8°C. For research use only.