Cation exchange chromatography columns, TSK-GEL® CM-5PW
Dodavatel: TOSOH Bioscience
Certifikáty
O tomto produktu
TSK-GEL® CM-5PW are polymer based stainless steel columns packed with ith porous hydrophilic polymer beads which are surface modified with carboxymethyl (CM) ligands to provide a weak cation exchange column. Used for the separation and analysis of proteins, peptides, and other biologically active molecules.
Frequently Bought Together