ContactUsLinkComponent
Moisture test kit, Hydranal™
Moisture test kit, Hydranal™
Catalog # 37858-1EA
CAS Number:  
Moisture test kit, Hydranal™
Catalog # 37858-1EA
CAS Number:  
Specifications
  • Typ balení:
    Glass bottle
  • Bal.:
    1
  • Popis:
    Moisture test kit
  • Environmentally Preferable:
Specifications
About this item
Test kit for the visual water determination according to Karl Fischer without titrator.