Zátkové chrániče sluchu, E-A-Rcap™
Catalog # 111-0912
Supplier: 3M
Specifications
- Barva:Modrá/žlutá
- Popis:Zátkové chrániče sluchu
- Environmentally Preferable:
- Bal.:50
Specifications
About this item
Banded earplugs are a practical option and easy to use.
- Extremely lightweight, virtually no feeling of pressure
- Plugs do not push into your ear canal; prevents irritation
- Ideal for frequent insertion and removal
They can be worn around the neck when not needed, making them ideal for temporary use.
SNR: 23 dB
Certifications: EN 352-2