Order Entry
Czech Republic
ContactUsLinkComponent
Anti-TSPAN5 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-TSPAN5 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Katalogové číslo BOSSBS-9414R-FITC
Dodavatel:  Bioss
Anti-TSPAN5 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Katalogové číslo BOSSBS-9414R-FITC
Dodavatel:  Bioss

Specifikace

  • Druh protilátky:
    Primární
  • Název antigenu:
    Tetraspanin 5
  • Symbol antigenu:
    TSPAN5
  • Klonovalita:
    Polyclonal
  • Spojení:
    FITC (Fluorescein Isothiocyanate)
  • Průtoková cytometrie:
    Yes
  • Hostitel:
    Rabbit
  • Imunochemie:
    Yes
  • Izotyp:
    IgG
  • Reaktivita:
    Human,
    Rat,
    Mouse
  • Křížová adsorbce:
    No
  • Formulář:
    liquid
  • Identifikační číslo genu:
    10098
  • Synonyma antigenu:
    tetraspanin 5|NET-4|tetraspan 5|transmembrane 4 superfamily member 8|TSPAN-5|transmembrane 4 superfamily member 9|tetraspanin-5|TM4SF9|tetraspan NET-4|tetraspan TM4SF
  • Skladovací pufr:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Skladovací teplota:
    Store at –20 °C for 12 months
  • Koncentrace:
    1 μg/μl
  • Transportní teplota:
    4°C
  • Čištění:
    Purified by Protein A
  • Bal.:
    100 µl

Specifikace

O tomto produktu

NET-4, also known as TSPAN5 or TM4SF9, is a 268 amino acid multi-pass membrane protein that belongs to the tetraspanin family and is thought to play a role in signal transduction events related to cell development, activation, growth and motility. The gene encoding NET-4 maps to human chromosome 4, which encodes nearly 6% of the human genome and has the largest gene deserts (regions of the genome with no protein encoding genes) of all of the human chromosomes. Defects in some of the genes located on chromosome 4 are associated with Huntington's disease, Ellis-van Creveld syndrome, methylmalonic acidemia and polycystic kidney disease.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: TSPAN5
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat