Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-CCP110 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
  BOSSBS-9307R-FITC
 :  Bioss
Anti-CCP110 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
  BOSSBS-9307R-FITC
 :  Bioss

 

  • Druh protilátky:
    Primární
  • Název antigenu:
    Centriolar Coiled Coil Protein 110kDa
  • Symbol antigenu:
    CCP110
  • Klonovalita:
    Polyclonal
  • Spojení:
    FITC (Fluorescein Isothiocyanate)
  • Hostitel:
    Rabbit
  • Imunochemie:
    Yes
  • Izotyp:
    IgG
  • Reaktivita:
    Human,
    Rat,
    Mouse
  • Křížová adsorbce:
    No
  • Formulář:
    liquid
  • Identifikační číslo genu:
    9738
  • Synonyma antigenu:
    Cep110|Centrosomal protein of 110 kDa|KIAA0419|CP110|CEP110|Centriolar coiled-coil protein of 110 kDa
  • Skladovací pufr:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Skladovací teplota:
    Store at –20 °C for 12 months
  • Koncentrace:
    1 μg/μl
  • Transportní teplota:
    4°C
  • Čištění:
    Purified by Protein A
  • Bal.:
    100 µl

 

 

Necessary for centrosome duplication at different stages of procentriole formation. Collaborates with CEP97, being involved in the suppression of a cilia assembly program. Required for correct spindle formation and has a role in regulating cytokinesis and genome stability via cooperation with CALM1 and CETN2.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CCP110
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat