Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-CATSPER4 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
  BOSSBS-6422R-A647
 :  Bioss
Anti-CATSPER4 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
  BOSSBS-6422R-A647
 :  Bioss

 

  • Druh protilátky:
    Primární
  • Název antigenu:
    Cation Channel, Sperm Associated 4
  • Symbol antigenu:
    CATSPER4
  • Klonovalita:
    Polyclonal
  • Spojení:
    Alexa Fluor® 647
  • Hostitel:
    Rabbit
  • Imunochemie:
    Yes
  • Izotyp:
    IgG
  • Reaktivita:
    Human
  • Křížová adsorbce:
    No
  • Formulář:
    liquid
  • Synonyma antigenu:
    sperm-associated 4.|CTSR4_HUMAN|CatSper4|Cation channel sperm-associated protein 4|CatSper-4|cation channel
  • Skladovací pufr:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Skladovací teplota:
    Store at 4°C for 12 months
  • Koncentrace:
    1 μg/μl
  • Transportní teplota:
    4°C
  • Čištění:
    Purified by Protein A
  • Bal.:
    100 µl

 

 

CatSpers (cation channel, sperm associated proteins) are ion transport proteins located on the surface of sperm cells in the principal piece of the sperm tail. CatSpers are vital to sperm motility, fertilization and cAMP-mediated calcium influx in sperm. There are four CatSper proteins in mammalian sperm, namely CatSper (or CatSper1), CatSper2, CatSper3 and CatSper4. CatSper proteins contain a single, six-transmembrane-spanning segment and exhibit the voltage-dependent Ca2+ channel four-repeat structure. CatSper proteins are believed to assemble into a heterotetrameric complex, forming an alkalinization-activated Ca2+-selective channel. Mutations in any of the genes encoding CatSper family proteins can result in male infertility. CatSper3 plays an important role in the hyperactivated motility of sperm cells, a process that is required in the preparation of sperm for fertilization.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CATSPER4
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human