Order Entry
Czech Republic
ContactUsLinkComponent
Anti-SYN1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anti-SYN1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Katalogové číslo BOSSBS-5631R-HRP
Dodavatel:  Bioss
Anti-SYN1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Katalogové číslo BOSSBS-5631R-HRP
Dodavatel:  Bioss

Specifikace

  • Druh protilátky:
    Primární
  • Název antigenu:
    Synapsin 1
  • Symbol antigenu:
    SYN1
  • Klonovalita:
    Polyclonal
  • Spojení:
    HRP (Horseradish Peroxidase)
  • Hostitel:
    Rabbit
  • Imunochemie:
    Yes
  • Izotyp:
    IgG
  • Reaktivita:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Křížová adsorbce:
    No
  • Formulář:
    liquid
  • Identifikační číslo genu:
    6853
  • Synonyma antigenu:
    SYN1a|Synapsin-1|Synapsin I|SYN1|Brain protein 4.1|SYNI|SYN1b
  • Modifikace:
    phospho-Ser603
  • Skladovací pufr:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
  • Skladovací teplota:
    Store at 4°C for 12 months
  • Koncentrace:
    1 μg/μl
  • Transportní teplota:
    4°C
  • Čištění:
    Purified by Protein A
  • Bal.:
    100 µl

Specifikace

O tomto produktu

Synapsin I is a member of the synapsin family. Synapsins are neuronal phosphoproteins which associate with the cytoplasmic surface of synaptic vesicles. Family members are characterized by common protein domains, and they are implicated in synaptogenesis and the modulation of neurotransmitter release, suggesting a potential role in several neuropsychiatric diseases. This member of the synapsin family plays a role in regulation of axonogenesis and synaptogenesis. The protein serves as a substrate for several different protein kinases and phosphorylation may function in the regulation of this protein in the nerve terminal. Mutations of the Synapsin I gene may be associated with X linked disorders with primary neuronal degeneration such as Rett syndrome.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: SYN1
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat