Order Entry
Czech Republic
ContactUsLinkComponent
Anti-MAGEA4 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-MAGEA4 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Katalogové číslo BOSSBS-3645R-FITC
Dodavatel:  Bioss
Anti-MAGEA4 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Katalogové číslo BOSSBS-3645R-FITC
Dodavatel:  Bioss

Specifikace

  • Druh protilátky:
    Primární
  • Název antigenu:
    Melanoma Antigen Family A4
  • Symbol antigenu:
    MAGEA4
  • Klonovalita:
    Polyclonal
  • Spojení:
    FITC (Fluorescein Isothiocyanate)
  • Hostitel:
    Rabbit
  • Imunochemie:
    Yes
  • Izotyp:
    IgG
  • Reaktivita:
    Human
  • Křížová adsorbce:
    No
  • Formulář:
    liquid
  • Identifikační číslo genu:
    4103
  • Synonyma antigenu:
    MAGE4A|CT1.4|MAGE4B|MAGE-X2|MAGE-41|MAGE4
  • Skladovací pufr:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Skladovací teplota:
    Store at 4°C for 12 months
  • Koncentrace:
    1 μg/μl
  • Transportní teplota:
    4°C
  • Čištění:
    Purified by Protein A
  • Bal.:
    100 µl

Specifikace

O tomto produktu

This gene is a member of the MAGEA gene family. The members of this family encode proteins with 50 to 80% sequence identity to each other. The promoters and first exons of the MAGEA genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. The MAGEA genes are clustered at chromosomal location Xq28. They have been implicated in some hereditary disorders, such as dyskeratosis congenita. At least four variants encoding the same protein have been found for this gene. [provided by RefSeq, Jul 2008].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MAGEA4
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human