Order Entry
Czech Republic
ContactUsLinkComponent
Anti-EPHB1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anti-EPHB1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # BOSSBS-10139R-HRP
Supplier:  Bioss
Anti-EPHB1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # BOSSBS-10139R-HRP
Supplier:  Bioss

Specifications

  • Druh protilátky:
    Primární
  • Název antigenu:
    EPH Receptor B1
  • Symbol antigenu:
    EPHB1
  • Klonovalita:
    Polyclonal
  • Spojení:
    HRP (Horseradish Peroxidase)
  • Hostitel:
    Rabbit
  • Imunochemie:
    Yes
  • Izotyp:
    IgG
  • Reaktivita:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Křížová adsorbce:
    No
  • Formulář:
    liquid
  • Identifikační číslo genu:
    2047
  • Synonyma antigenu:
    Ephrin type-B receptor 2|EphrinB1|Ephrin type-B receptor 3|HEK2|Hek5|NET|Ephrin B3|CAPB|ERK|EPHT3|EphrinB3|PCBC|ELK|Ephrin type-B receptor 1|Ephrin B2|TYRO6|HEK-6|EphrinB2|Tyrosine-protein kinase receptor EPH-2|Tyro5|Ephrin B1|ETK2|MGC87492|EPHB2|EPHB3|DRT
  • Modifikace:
    phospho-Tyr968
  • Skladovací pufr:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
  • Skladovací teplota:
    Store at 4°C for 12 months
  • Koncentrace:
    1 μg/μl
  • Transportní teplota:
    4°C
  • Čištění:
    Purified by Protein A
  • Bal.:
    100 µl

Specifications

About this item

EphB1, previously known as Elk (eph like kinase), is a receptor tyrosine kinase of the highly tissue restricted family of eph proteins. EphB1 and other ephB family members are type 1 membrane spanning proteins, comprised of immunoglobulin, fibronectin type III, and cysteine rich subdomains in the ecto domain, and the single uninterrupted cytoplasmic tyrosine kinase domain upstream of a carboxyterminal sterile alpha motif (SAM) domain. EphB family proteins bind ephrins of the B class. EphB1 is expressed predominately in developing neural structures in embryos, and in vascular epithelium of kidney, and other tissues. Upon binding to alternatively oligomerized ephrin B1, EphB1 signals regulation of cell attachment and cell to cell assembly. Members of this protein family are implicated in neuronal and vascular cell targeting.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: EPHB1
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat