Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-UBE2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
  BOSSBS-3814R-FITC
 :  Bioss
Anti-UBE2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
  BOSSBS-3814R-FITC
 :  Bioss

 

  • Druh protilátky:
    Primární
  • Název antigenu:
    Ubiquitin-activating Enzyme 2
  • Symbol antigenu:
    UBE2
  • Klonovalita:
    Polyclonal
  • Spojení:
    FITC (Fluorescein Isothiocyanate)
  • Hostitel:
    Rabbit
  • Imunochemie:
    Yes
  • Izotyp:
    IgG
  • Reaktivita:
    Human,
    Rat,
    Mouse
  • Křížová adsorbce:
    No
  • Formulář:
    liquid
  • Identifikační číslo genu:
    7318
  • Synonyma antigenu:
    Ube1L
  • Skladovací pufr:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Skladovací teplota:
    Store at 4°C for 12 months
  • Koncentrace:
    1 μg/μl
  • Transportní teplota:
    4°C
  • Čištění:
    Purified by Protein A
  • Bal.:
    100 µl

 

 

The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E1 ubiquitin-activating enzyme family. The encoded enzyme is a retinoid target that triggers promyelocytic leukemia (PML)/retinoic acid receptor alpha (RARalpha) degradation and apoptosis in acute promyelocytic leukemia, where it is involved in the conjugation of the ubiquitin-like interferon-stimulated gene 15 protein. [provided by RefSeq, Jul 2008].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: UBE2
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat