Order Entry
Czech Republic
ContactUsLinkComponent
Anti-PLB Rabbit Polyclonal Antibody (Cy7®)
Anti-PLB Rabbit Polyclonal Antibody (Cy7®)
  BOSSBS-2984R-CY7
 :  Bioss
Anti-PLB Rabbit Polyclonal Antibody (Cy7®)
  BOSSBS-2984R-CY7
 :  Bioss

 

  • Druh protilátky:
    Primární
  • Název antigenu:
    Phospholamban
  • Symbol antigenu:
    PLB
  • Klonovalita:
    Polyclonal
  • Spojení:
    Cy7®
  • Hostitel:
    Rabbit
  • Imunochemie:
    Yes
  • Izotyp:
    IgG
  • Reaktivita:
    Human,
    Rat,
    Mouse
  • Křížová adsorbce:
    No
  • Formulář:
    liquid
  • Identifikační číslo genu:
    5350
  • Synonyma antigenu:
    PPLA_HUMAN.|PLN|Cardiac phospholamban|CMD1P|Phospholamban phospho S16|PLB|p-Phospholamban phospho S16|p-PLBS16
  • Modifikace:
    phospho-Ser16
  • Skladovací pufr:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Skladovací teplota:
    Store at 4°C for 12 months
  • Koncentrace:
    1 μg/μl
  • Transportní teplota:
    4°C
  • Čištění:
    Purified by Protein A
  • Bal.:
    100 µl

 

 

Defects in PLN are the cause of cardiomyopathy dilated type 1P (CMD1P) . Dilated cardiomyopathy is a disorder characterized by ventricular dilation and impaired systolic function, resulting in congestive heart failure and arrhythmia. Patients are at risk of premature death.Defects in PLN are the cause of cardiomyopathy familial hypertrophic type 18 (CMH18) . CMH18 is a hereditary heart disorder characterized by ventricular hypertrophy, which is usually asymmetric and often involves the interventricular septum. The symptoms include dyspnea, syncope, collapse, palpitations, and chest pain. They can be readily provoked by exercise. The disorder has inter- and intrafamilial variability ranging from benign to malignant forms with high risk of cardiac failure and sudden cardiac death.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PLB
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat