Order Entry
Czech Republic
ContactUsLinkComponent
Anti-CRX Rabbit Polyclonal Antibody (Cy7®)
Anti-CRX Rabbit Polyclonal Antibody (Cy7®)
Katalogové číslo BOSSBS-3798R-CY7
Dodavatel:  Bioss
Anti-CRX Rabbit Polyclonal Antibody (Cy7®)
Katalogové číslo BOSSBS-3798R-CY7
Dodavatel:  Bioss

Specifikace

  • Druh protilátky:
    Primární
  • Název antigenu:
    Cone-rod Homeobox
  • Symbol antigenu:
    CRX
  • Klonovalita:
    Polyclonal
  • Spojení:
    Cy7®
  • Hostitel:
    Rabbit
  • Imunochemie:
    Yes
  • Izotyp:
    IgG
  • Reaktivita:
    Human,
    Rat,
    Mouse
  • Křížová adsorbce:
    No
  • Formulář:
    liquid
  • Identifikační číslo genu:
    1406
  • Synonyma antigenu:
    LCA7|OTX3|CRD|CORD2
  • Skladovací pufr:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Skladovací teplota:
    Store at 4°C for 12 months
  • Koncentrace:
    1 μg/μl
  • Transportní teplota:
    4°C
  • Imunogen:
    60-110/299
  • Čištění:
    Purified by Protein A
  • Bal.:
    100 µl

Specifikace

O tomto produktu

Transcription factor that binds and transactivates the sequence 5'-TAATC[CA]-3' which is found upstream of several photoreceptor-specific genes, including the opsin genes. Acts synergistically with other transcription factors, such as NRL, RORB and RAX, to regulate photoreceptor cell-specific gene transcription. Essential for the maintenance of mammalian photoreceptors.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CRX
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype:
Reactivity: