Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-ADM Rabbit Polyclonal Antibody (Cy3®)
  BOSSBS-0995R-CY3
 :  Bioss
Anti-ADM Rabbit Polyclonal Antibody (Cy3®)
  BOSSBS-0995R-CY3
 :  Bioss

 

  • Druh protilátky:
    Primární
  • Název antigenu:
    Adrenomedullin
  • Symbol antigenu:
    ADM
  • Klonovalita:
    Polyclonal
  • Spojení:
    Cy3®
  • Hostitel:
    Rabbit
  • Imunochemie:
    Yes
  • Izotyp:
    IgG
  • Reaktivita:
    Human,
    Rat,
    Mouse
  • Křížová adsorbce:
    No
  • Formulář:
    liquid
  • Identifikační číslo genu:
    133
  • Synonyma antigenu:
    AM|Proadrenomedullin N-20 terminal peptide|ADM|PAMP|ProAM N-terminal 20 peptide|ProAM-N20
  • Skladovací pufr:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Skladovací teplota:
    Store at 4°C for 12 months
  • Koncentrace:
    1 μg/μl
  • Transportní teplota:
    4°C
  • Čištění:
    Purified by Protein A
  • Bal.:
    100 µl

 

 

Adrenomedullin (ADM), a vasodilator produced by most contractile cells, is characterized by persistent hypotensive activity. ADM is involved in the regulation of fluid and electrolyte homeostasis and in the maintenance of cardiovascular functioning. In hypertensive patients, the level of ADM in plasma is up-regulated. Natriuresis is a common systemic manifestation of aneurysmal subarachnoid hemorrhage. ADM has strong natriuretic actions. ADM-induced natriuresis is caused by an increase in glomerular filtration rate and a decrease in distal tubular sodium reabsorption. ADM is present both in the periphery and brain, and can exert central effects such as decreasing food ingestion.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ADM
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat