Specifikace
- Druh protilátky:Primární
- Název antigenu:DNA Cross-link Repair 1C
- Symbol antigenu:DCLRE1C
- Klonovalita:Polyclonal
- Spojení:Cy7®
- Hostitel:Rabbit
- Imunochemie:Yes
- Izotyp:IgG
- Reaktivita:Human,Rat,Mouse
- Křížová adsorbce:No
- Formulář:liquid
- Identifikační číslo genu:64421
- Synonyma antigenu:SNM1C|A-SCID|RS-SCID|SCIDA|DCLREC1C
- Skladovací pufr:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Skladovací teplota:Store at 4°C for 12 months
- Koncentrace:1 μg/μl
- Transportní teplota:4°C
- Imunogen:350-400/692
- Čištění:Purified by Protein A
- Bal.:100 µl
Specifikace
O tomto produktu
Required for V(D)J recombination, the process by which exons encoding the antigen-binding domains of immunoglobulins and T-cell receptor proteins are assembled from individual V, (D), and J gene segments. V(D)J recombination is initiated by the lymphoid specific RAG endonuclease complex, which generates site specific DNA double strand breaks (DSBs). These DSBs present two types of DNA end structures: hairpin sealed coding ends and phosphorylated blunt signal ends. These ends are independently repaired by the non homologous end joining (NHEJ) pathway to form coding and signal joints respectively. This protein exhibits single-strand specific 5'-3' exonuclease activity in isolation and acquires endonucleolytic activity on 5' and 3' hairpins and overhangs when in a complex with PRKDC. The latter activity is required specifically for the resolution of closed hairpins prior to the formation of the coding joint. May also be required for the repair of complex DSBs induced by ionizing radiation, which require substantial end-processing prior to religation by NHEJ.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: DCLRE1C
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat