Order Entry
Czech Republic
ContactUsLinkComponent
Anti-FUT11 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anti-FUT11 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-12259R-A488
Supplier:  Bioss
Anti-FUT11 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-12259R-A488
Supplier:  Bioss

Specifications

  • Druh protilátky:
    Primární
  • Název antigenu:
    Fucosyltransferase 11
  • Symbol antigenu:
    FUT11
  • Klonovalita:
    Polyclonal
  • Spojení:
    Alexa Fluor® 488
  • Hostitel:
    Rabbit
  • Imunochemie:
    Yes
  • Izotyp:
    IgG
  • Reaktivita:
    Human,
    Rat,
    Mouse
  • Křížová adsorbce:
    No
  • Formulář:
    liquid
  • Identifikační číslo genu:
    170384
  • Skladovací pufr:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Skladovací teplota:
    Store at 4°C for 12 months
  • Koncentrace:
    1 μg/μl
  • Transportní teplota:
    4°C
  • Čištění:
    Purified by Protein A
  • Bal.:
    100 µl

Specifications

About this item

Glycosyltransferases that mediate the regio- and stereoselective transfer of sugars, such as the fucosyltransferases, determine cell surface-carbohydrate profiles, which are essential interfaces for biological recognition processes. Fucosyltransferases (FucTs) catalyze the covalent association of fucose to different positional linkages on sugar acceptor molecules. The carbohydrate moieties that are generated are covalently attached to cell surfaces and are necessary to ensure a surface contour that satisfies a variety of physiological roles. FucT-XI is a 492 amino acid single-pass type II membrane protein that belongs to the glycosyltransferase 10 family. Localizing to Golgi apparatus, FucT-XI may act as a fucosyltransferase and exists as two alternatively spliced isoforms. The gene encoding FucT-XI maps to mouse chromosome 14 A3.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FUT11/FucT-XI
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat