Order Entry
Czech Republic
ContactUsLinkComponent
Anti-SCN2B Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anti-SCN2B Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Katalogové číslo BOSSBS-12122R-A488
Supplier:  Bioss
Anti-SCN2B Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Katalogové číslo BOSSBS-12122R-A488
Supplier:  Bioss

Specifications

  • Druh protilátky:
    Primární
  • Název antigenu:
    Sodium Channel Voltage-gated Type II Beta
  • Symbol antigenu:
    SCN2B
  • Klonovalita:
    Polyclonal
  • Spojení:
    Alexa Fluor® 488
  • Průtoková cytometrie:
    Yes
  • Hostitel:
    Rabbit
  • Imunochemie:
    Yes
  • Izotyp:
    IgG
  • Reaktivita:
    Human,
    Rat,
    Mouse
  • Křížová adsorbce:
    No
  • Formulář:
    liquid
  • Synonyma antigenu:
    SCNB2|ATFB14
  • Skladovací pufr:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Skladovací teplota:
    Store at 4°C for 12 months
  • Koncentrace:
    1 μg/μl
  • Transportní teplota:
    4°C
  • Čištění:
    Purified by Protein A
  • Bal.:
    100 µl

Specifications

About this item

Voltage-gated sodium channels are selective ion channels that regulate the permeability of sodium ions in excitable cells. During the propagation of an action potential, sodium channels allow an influx of sodium ions, which rapidly depolarizes the cell. Na+ CP type II beta(sodium channel, voltage-gated, type II, beta), also known as SCN2B, is a 215 amino acid single-pass type I membrane protein that plays a critical role in the expression and assembly of the heterotrimeric complex of the sodium channel and interacts with Tenascin-R to influence the clustering and regulation of sodium channels at nodes of Ranvier. Expressed specifically in brain, Na+ CP type II beta contains one Ig-like C2-type (immunoglobulin-like) domain and is encoded by a gene that maps to human chromosome 11q23.3 and mouse chromosome 9 A5.2.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: SCN2B
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat