Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-SP7 Rabbit Polyclonal Antibody (Cy3®)
  BOSSBS-1110R-CY3
 :  Bioss
Anti-SP7 Rabbit Polyclonal Antibody (Cy3®)
  BOSSBS-1110R-CY3
 :  Bioss

 

  • Druh protilátky:
    Primární
  • Název antigenu:
    Transcription factor Sp7
  • Symbol antigenu:
    SP7
  • Klonovalita:
    Polyclonal
  • Spojení:
    Cy3®
  • Hostitel:
    Rabbit
  • Imunochemie:
    Yes
  • Izotyp:
    IgG
  • Reaktivita:
    Human,
    Rat,
    Mouse
  • Křížová adsorbce:
    No
  • Formulář:
    liquid
  • Identifikační číslo genu:
    121340
  • Synonyma antigenu:
    Osterix
  • Skladovací pufr:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Skladovací teplota:
    Store at 4°C for 12 months
  • Koncentrace:
    1 μg/μl
  • Transportní teplota:
    4°C
  • Imunogen:
    340-400/431
  • Čištění:
    Purified by Protein A
  • Bal.:
    100 µl

 

 

Transcriptional activator essential for osteoblast differentiation. Binds to SP1 and EKLF consensus sequences and to other G/C-rich sequences (By similarity).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SP7
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat