Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-PSEN1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Katalogové číslo BOSSBS-0024R-A488
Supplier:  Bioss
Anti-PSEN1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Katalogové číslo BOSSBS-0024R-A488
Supplier:  Bioss

Specifications

  • Druh protilátky:
    Primární
  • Název antigenu:
    Presenilin 1
  • Symbol antigenu:
    PSEN1
  • Klonovalita:
    Polyclonal
  • Spojení:
    Alexa Fluor® 488
  • Hostitel:
    Rabbit
  • Imunochemie:
    Yes
  • Izotyp:
    IgG
  • Reaktivita:
    Human,
    Rat,
    Mouse
  • Křížová adsorbce:
    No
  • Formulář:
    liquid
  • Identifikační číslo genu:
    5663
  • Synonyma antigenu:
    FAD|Alzheimer Disease 3|Homo Sapiens Clone CC44 Senilin 1|Senilin 1|PS1|PSNL1|AD3|S182 Protein
  • Skladovací pufr:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Skladovací teplota:
    Store at 4°C for 12 months
  • Koncentrace:
    1 μg/μl
  • Transportní teplota:
    4°C
  • Čištění:
    Purified by Protein A
  • Bal.:
    100 µl

Specifications

About this item

Alzheimer's disease (AD) patients with an inherited form of the disease carry mutations in the presenilin proteins (PSEN1; PSEN2) or in the amyloid precursor protein (APP). These disease-linked mutations result in increased production of the longer form of amyloid-beta (main component of amyloid deposits found in AD brains). Presenilins are postulated to regulate APP processing through their effects on gamma-secretase, an enzyme that cleaves APP. Also, it is thought that the presenilins are involved in the cleavage of the Notch receptor, such that they either directly regulate gamma-secretase activity or themselves are protease enzymes. Several alternatively spliced transcript variants encoding different isoforms have been identified for this gene, the full-length nature of only some have been determined. [provided by RefSeq, Aug 2008]

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PSEN1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat