ContactUsLinkComponent
Detergent, neodisher LaboClean FT
Catalog # WEIG410030_U
Supplier:  Dr. Weigert
undefined
Detergent, neodisher LaboClean FT
Catalog # WEIG410030_U
Supplier:  Dr. Weigert
Specifications
  • Popis:
    neodisher® LaboClean FT
  • Sekvence:
    02
  • Typ balení:
    Canister
  • Bal.:
    1
  • Teplotní rozsah:
    Store at 0...25 °C
  • pH rozsah:
    11,8 - 12,2 (3 - 4 ml/L, determined in deionised water, 20 °C)
Specifications
About this item
neodisher® LaboClean FT
  • Alkaline liquid concentrate especially suitable for removal of residues of culture media, blood, proteins and other organic materials
  • Components: Potassium hydroxide and sodium hypochlorite
  • Dosage: 3 to 4 ml/L
Caution: Most of the materials are corrosive or irritant and care must be taken with the choice of materials for cleaning aluminium and titanium items. Please read the Material Safety Data sheet before use.