Reference standards for food analysis, FAME mixtures, PUFA, Supelco®
Katalogové číslo 47033.
Supplier: Merck
CAS Number:
Specifications
- Bal.:100 mg
- Popis:PUFA No.1 (marine source)
- Sekvence:01
Specifications
About this item
Complex qualitative polyunsaturated fatty acid (PUFA) methyl esters standard mixtures, extracted from natural materials.