Anti-p-MEK1 Rabbit Polyclonal Antibody [clone: S298]
Katalogové číslo BWRLAP0092
Dodavatel: Bioworld Technology
Specifikace
- Druh protilátky:Primární
- Symbol antigenu:p-MEK1
- Klonovalita:Polyclonal
- Klon:S298
- Hostitel:Rabbit
- Reaktivita:Human,Rat,Mouse
- Western blot:Yes
- Rozměry protilátky:100 µg
- Bal.:100 µG
Specifikace
O tomto produktu
Synthetic phosphopeptide derived from human MEK1 around the phosphorylation site of Serine 298.
p-MEK1(S298) pAb detects endogenous levels of MEK1 protein only when phosphorylated at Ser298.
Type: Primary
Antigen: p-MEK1
Clonality: Polyclonal
Clone: S298
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat