Anti-MEK1 Rabbit Polyclonal Antibody [clone: P293]
Catalog # BWRLBS3246
Supplier: Bioworld Technology
Specifications
- Druh protilátky:Primární
- Symbol antigenu:MEK1
- Klonovalita:Polyclonal
- Klon:P293
- Hostitel:Rabbit
- Imunochemie:Yes
- Reaktivita:Human,Rat,Mouse
- Western blot:Yes
- Rozměry protilátky:100 µg
- Bal.:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 270-320 of Human MEK1.
MEK1 (P293) pAb detects endogenous levels of MEK1 protein.
Type: Primary
Antigen: MEK1
Clonality: Polyclonal
Clone: P293
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat