Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-MEK1/2 Rabbit Polyclonal Antibody [clone: S211]
Catalog # BWRLBS3599
Anti-MEK1/2 Rabbit Polyclonal Antibody [clone: S211]
Catalog # BWRLBS3599

Specifications

  • Druh protilátky:
    Primární
  • Symbol antigenu:
    MEK1/2
  • Klonovalita:
    Polyclonal
  • Klon:
    S211
  • Hostitel:
    Rabbit
  • Reaktivita:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Rozměry protilátky:
    100 µg
  • Bal.:
    100 µG

Specifications

About this item

Synthetic peptide, corresponding to amino acids 180-230 of Human MEK1.

MEK1/2 (S211) pAb detects endogenous levels of MEK1/2 protein.

Type: Primary
Antigen: MEK1/2
Clonality: Polyclonal
Clone: S211
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat