Anti-Na+ CP-pan Rabbit Polyclonal Antibody [clone: K1493]
Catalog # BWRLBS1328
Supplier: Bioworld Technology
Specifications
- Druh protilátky:Primární
- Symbol antigenu:Na+ CP-pan
- Klonovalita:Polyclonal
- Klon:K1493
- Hostitel:Rabbit
- Imunochemie:Yes
- ImunoFluorescence:Yes
- Reaktivita:Human,Rat,Mouse
- Western blot:Yes
- Rozměry protilátky:100 µg
- Bal.:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 1470-1520 of Human SCN5A.
Sodium Channel-pan (K1493) pAb detects endogenous levels of Sodium Channel-pan protein.
Type: Primary
Antigen: Na+ CP-pan
Clonality: Polyclonal
Clone: K1493
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat