Extractant for nitrate and nitrogen in soils, calcium sulphate powder
Catalog # HACH1232120
Supplier: Hach
CAS Number:
Specifications
- Bal.:20 g
- Popis:Extractant for nitrate and nitrogen in soils, calcium sulphate powder
- Testů v kitu:50
Specifications
About this item
This calcium sulphate extractant is used for nitrate and nitrogen determination in soils.