Shorting strap for BNC meters, Orion™
Catalog # ORIO090045
Supplier: Thermo Orion
Specifications
- Popis:BNC meter shorting strap for pH/ISE/conductivity/RDO/DO meters with BNC input
- Sekvence:110
- Bal.:1
Specifications
About this item
Shorting strap for use on pH meters with BNC input to perform BNC input verification testing and meter self test.
For use with: pH meters with BNC input (Orion Versa Star pH, pH/ISE and pH/LogR meters; Orion Star A series pH, pH/ISE, pH/conductivity, pH/RDO/DO and pH/ISE/conductivity/RDO/DO meters; Orion Dual Star pH/ISE meters; Orion 2-Star, 3-Star, 4-Star and 5-Star pH meters (retired)).