Orders LinkContactUsLinkComponent
FlashGel™ RNA system
FlashGel™ RNA system
Dodavatel:   LONZA
FlashGel™ RNA system
Dodavatel:  LONZA
Certifikáty
FlashGel™ RNA system
3 Dostupných variant níže
Product Details & Documents
FlashGel™ system for RNA is optimised for the unique requirements of RNA, and is an ideal tool for rapid analysis of sample integrity. High quality, intact RNA is essential for consistent results in gene expression, Northern analysis, cDNA library construction and cDNA labelling for microarrays. Most protocols recommend checking RNA prior to downstream analysis.
  • Complete RNA analysis in less than 30 minutes
  • Detects <10 ng RNA per band, separation range 0,5 to 9 Kb
  • Avoids the use of hazardous reagents and contaminating RNases
Certifications: Some components and technology of the FlashGel™ system are sold under licensing agreements. The nucleic acid stain in this product is manufactured and sold under license from Molecular Probes, Inc for use only in research applications or quality control, and is covered by pending and issued patents. The FlashGel Dock technology is licensed from Clare Chemical Research, Inc. The electrophoresis technology is licensed from Temple University.
Informace o dodávce: Supplied as separate components or as a starter kit. Starter kit contains box of 9, 1,2%, 12+1 well FlashGel™ RNA cassettes; sample buffer; RNA marker; and molecular biology grade water. FlashGel™ dock sold separately.
Upozornení: Gel cannot be accessed. Unsuitable for recovery.
Specifications for Product Family
Specifikace
  • Catalog No:
  • 733-1612
  • 733-1617
  • 733-1618
  • Model:
  • FlashGel™ RNA
  • FlashGel™ RNA
  • FlashGel™ RNA
  • Velikost gelu Š x D:
  • 84×70 mm
  • 84×70 mm
  • 84×70 mm
  • Rozsah dělení (bp):
  • 0,5 kb - 9 kb
  • 0,5 kb - 9 kb
  • 0,5 kb - 9 kb
  • Uskladnění (kazety):
  • Room temperature for 3 months from date of manufacture
  • Room temperature for 3 months from date of manufacture
  • Room temperature for 3 months from date of manufacture
  • Objem jamky:
  • 2 - 5 µl
  • 2 - 5 µl
  • 2 - 5 µl
  • Velikost kazety ŠxHxV:
  • 107×115×17 mm
  • 107×115×17 mm
  • 107×115×17 mm
  • Velikost okna ŠxH×V:
  • 120×134×54 mm
  • 120×134×54 mm
  • 120×134×54 mm
Specifikace
Product Family Options
Informace o produktu
  • Sekvence
    Dostupnost
    Price
  • 733-1612
    FlashGel™ system for RNA, starter kit (without dock)
    FlashGel™ system for RNA, starter kit (without dock)
    03
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KIT
    1 * 1 KIT - 17 390,00 Kč
    17 390,00 Kč
    1 * 1 KIT
       17 390,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    733-1612
    Model
    FlashGel™ RNA
    Velikost gelu Š x D
    84×70 mm
    Rozsah dělení (bp)
    0,5 kb - 9 kb
    Uskladnění (kazety)
    Room temperature for 3 months from date of manufacture
    Objem jamky
    2 - 5 µl
    Velikost kazety ŠxHxV
    107×115×17 mm
    Velikost okna ŠxH×V
    120×134×54 mm
  • 733-1617
    FlashGel™ RNA cassettes, 1,2% agarose, 12+1 single tier
    FlashGel™ RNA cassettes, 1,2% agarose, 12+1 single tier
    09
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 9 KS
    1 * 9 KS - 6 060,00 Kč
    6 060,00 Kč
    1 * 9 KS
       6 060,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    733-1617
    Model
    FlashGel™ RNA
    Velikost gelu Š x D
    84×70 mm
    Rozsah dělení (bp)
    0,5 kb - 9 kb
    Uskladnění (kazety)
    Room temperature for 3 months from date of manufacture
    Objem jamky
    2 - 5 µl
    Velikost kazety ŠxHxV
    107×115×17 mm
    Velikost okna ŠxH×V
    120×134×54 mm
  • 733-1618
    FlashGel™ RNA cassettes, 1,2% agarose, 16+1 double tier
    FlashGel™ RNA cassettes, 1,2% agarose, 16+1 double tier
    10
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 9 KS
    1 * 9 KS - 6 060,00 Kč
    6 060,00 Kč
    1 * 9 KS
       6 060,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    733-1618
    Model
    FlashGel™ RNA
    Velikost gelu Š x D
    84×70 mm
    Rozsah dělení (bp)
    0,5 kb - 9 kb
    Uskladnění (kazety)
    Room temperature for 3 months from date of manufacture
    Objem jamky
    2 - 5 µl
    Velikost kazety ŠxHxV
    107×115×17 mm
    Velikost okna ŠxH×V
    120×134×54 mm
Accessories
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • FlashGel™ power supply
    FlashGel™ power supply
    LONZ57068
    12
    Přihlásit se pro dostupnost
    1 * 1 KIT - 22 090,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • FlashGel™ power supply pack, includes FlashGel™ dock
    FlashGel™ power supply pack, includes FlashGel™ dock
    LONZ57069
    13
    Přihlásit se pro dostupnost
    1 * 1 KIT - 43 490,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • FlashGel™ dock
    FlashGel™ dock
    733-1609
    14
    Přihlásit se pro dostupnost
    1 * 1 KS - 24 090,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • RNA marker (0,5 bp to 9 Kb)
    RNA marker (0,5 bp to 9 Kb)
    733-1734
    19
    Přihlásit se pro dostupnost
    1 * 50 µG - 14 060,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • Formaldehyde sample buffer
    Formaldehyde sample buffer
    733-1739
    23
    Přihlásit se pro dostupnost
    1 * 5 mL - 2 140,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • RNA marker (0,5 - 9 kb), 50 µL
    RNA marker (0,5 - 9 kb), 50 µL
    733-1623
    22
    Přihlásit se pro dostupnost
    1 * 50 µl - 8 280,00 Kč