Benchmate vial rack, CryoELITE®, WHEATON®
Catalog # 479-2111
Supplier: DWK Life Sciences
Specifications
- Popis:Benchmate rack
- Bal.:5
Specifications
About this item
The CryoELITE® benchmate rack can be used for both freestanding and round bottom vials. The Loctagon™ bottom allows for the one hand removal of freestanding vial caps.
Holds up to 50 cryogenic vials. Manufactured from polypropylene, the rack is easily celaned in an automatic washer or autoclavable at 121 °C for 20 minutes. Non-skid feet offer additional stability for bench work. Each well has an inner diameter of 12,5 mm and the rack has an overall footprint of 190×100×22 mm. CryoELITE® benchmate vial rack is stackable. The freestanding vials will lock into the rack.