UHPLC precolumn filters, UltraShield
Catalog # REST25810
Supplier: Restek
Specifications
- Materiál frity:Titanium
- Porozita:0,5 µm
- Popis:Ultrashield UHPLC Precolumn filter with female/male 10-32 connections, 0,2 μm frit
- Sekvence:04
- Bal.:1
Specifications
About this item
Titanium filter in a stainless steel body with PEEK ferrule. Its design easily fits with all columns.
- Economical protection for UHPLC systems
- Reliable way to extend column lifetime
- Pressure rated to 15000 psi (1,034 bar)
Inlet/outlet connection: Female/male 10 to 32
Port geometry: Parker (1/16 CPI)
Wrench flat: ⁵/₁₆"