Inlet liners for Thermo Scientific GCs
Katalogové číslo REST22288
Dodavatel: Restek
Specifikace
- Ø vněj.:8,0 mm
- Ø vnitř.:5,0 mm
- Type:For Thermo TRACE, 8000 Series and Focus GCs
- Popis:Split precision liner with deactivated wool, borosilicate glass, intermediate polarity deactivated, 1/pack
- Délka:105 mm
- Sekvence:76
- Bal.:1
Specifikace
O tomto produktu
Use these liners for increased sensitivity. They are manufactured from borosilicate glass or metal.
Int.Ø=Nominal diameter at syringe needle expulsion point.