Orders LinkContactUsLinkComponent
Dissecting scissors, type Kelly
Dissecting scissors, type Kelly
03, Kelly, curved, saw edge, sharp/sharp, 160 mm
Catalog # HAMMHSB033-16
Supplier:   HAMMACHER KARL
Dissecting scissors, type Kelly
Catalog # HAMMHSB033-16
Supplier:  HAMMACHER KARL
Dissecting scissors, type Kelly
Dissecting scissors, type Kelly
03, Kelly, curved, saw edge, sharp/sharp, 160 mm
Price
2 620,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Dissecting scissors made of stainless steel.
Specifications
  • Type:
    Kelly, curved, saw edge, sharp/sharp
  • Délka:
    160 mm
  • Sekvence:
    03