Anti-MUT Rabbit Polyclonal Antibody
Katalogové číslo BSBTA01065
Dodavatel: Boster Bio
Specifikace
- Druh protilátky:Primární
- Symbol antigenu:MUT
- Klonovalita:Polyclonal
- Hostitel:Rabbit
- Imunochemie:Yes
- Izotyp:IgG
- Reaktivita:Human,Rat
- Western blot:Yes
- Rozměry protilátky:100 µg
- Bal.:100 µG
Specifikace
O tomto produktu
Rabbit IgG polyclonal antibody for Methylmalonyl-CoA mutase, mitochondrial(MUT) detection. Tested with WB, IHC-P in Human, Rat.
Type: Primary
Antigen: MUT
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat