Anti-HLA-DPB1 Rabbit Polyclonal Antibody
Catalog # BSBTA00487
Supplier: Boster Bio
Specifications
- Druh protilátky:Primární
- Symbol antigenu:HLA-DPB1
- Klonovalita:Polyclonal
- Hostitel:Rabbit
- Imunochemie:Yes
- Izotyp:IgG
- Reaktivita:Human,Rat,Mouse
- Western blot:Yes
- Rozměry protilátky:100 µg
- Bal.:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for HLA class II histocompatibility antigen, DP beta 1 chain (HLA-DPB1) detection. Tested with WB in Human, Mouse, Rat.
Type: Primary
Antigen: HLA-DPB1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat