Hlavy peristalitického čerpadla, rotarus®
Supplier: HIRSCHMANN
Certificates
About this item
Specifications
- Catalog No:
- 224-0123
- 224-0135
- 224-0121
- 224-0122
- 224-0134
- Model:
- MKF 12-8
- PK 10-24
- MKF 4-8
- MKF 8-4
- PK 10-16
- Minimální průtok:
- 0,0001 ml/min
- 0,66 ml/min
- 0,0009 ml/min
- 0,014 ml/min
- 0,08 ml/min
- Počet válečků:
- 8
- 3
- 8
- 4
- 3
Specifications
Related Information
Hirschmann brochure insert sheets rotarus® -pump series- tubes Hirschmann brochure rotarus® -pump series-
Product Literature
Hirschmann data sheets rotarus® peristaltic pump head MKF12-8 Hirschmann data sheets rotarus® peristaltic pump head MKF4-8 Hirschmann data sheets rotarus® peristaltic pump head MKF8-4 Hirschmann data sheets rotarus® peristaltic pump head PK10-16 Hirschmann data sheets rotarus® peristaltic pump head PK10-24
Datasheet
Frequently Bought Together