Calf collagen (from skin), MP Biomedicals
Dodavatel: MP Biomedicals
Certifikáty
O tomto produktu
Collagen is a group of naturally occurring proteins found in animals, especially in the flesh and connective tissues of vertebrates. It is a component of skin, bone, tendon, and other fibrous connective tissues.
- Soluble in 0,075 M Sodium citrate and in dilute acetic acid
- Suitable for gel formation, platelet aggregation, collagenase activity
Specifikace
- Čí. CAS:9007-34-5
- Zdroj:Skin
- Druhy:Calf
- Podmínky uskladnění:+4 °C
- Název proteinu/peptidu:Collagen
- Barva:White
- Formulace:Lyophilised fluffy solid