Capillary columns, Rxi®-XLB (fused silica)
Supplier: Restek
Certificates
About this item
Rxi®-XLB (fused silica) columns are ideal for many GC-MS applications, including pesticides, PCB congeners (e.g., Aroclor mixes), PAHs.
- Ultra-low bleed
- Unique selectivity
- Temperature range: 30 to 360 °C
Ideal for analysing active, high molecular weight compounds. It is coupled with inertness and high thermal stability, make Rxi-XLB columns ideal for analysing semivolatile compounds in drinking water.
Frequently Bought Together