Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Meřiče elektrochemické
Měřiče rozpušteného kyslíku
Membrane kit for dissolved oxygen meter, YSI 55
Membrane kit for dissolved oxygen meter, YSI 55
Membrane kit for dissolved oxygen meter, YSI 55
YSIA098094
:
YSI ENVIRONMENTAL
Membrane kit for dissolved oxygen meter, YSI 55
YSIA098094
:
YSI ENVIRONMENTAL
Popis:
Standard membrane kit for polarographic probes, incl. KCL electrolyte (1 mil)
Bal.:
1