Order Entry
Czech Republic
ContactUsLinkComponent
Recombinant CPN-Ag PmpD
Recombinant CPN-Ag PmpD
Catalog # BIRBORB98899-1
Supplier:  Biorbyt
CAS Number:  
Recombinant CPN-Ag PmpD
Catalog # BIRBORB98899-1
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Bal.:
    1 mg
  • Typ proteinu/peptidu:
    Recombinant
  • Podmínky uskladnění:
    Store at −20 °C. Avoid multiple freeze/thaw cycles.
  • Název proteinu/peptidu:
    CPN-Ag PmpD
  • Čistota:
    Greater than 95% pure SDS PAGE non-reducing, Electrophoresis
  • Formulace:
    PBS, pH 7.4
  • Testované aplikace:
    ELISA , WB , IHC-P , RIA

Specifications

About this item