Leptin
BIRBORB97815-100
: Biorbyt
:
- Bal.:100 µG
- Podmínky uskladnění:Store at−20 °C.
- Biologická aktivita:This Leptin triple mutant is capable of inhibiting leptininduced proliferation of BAF/3 cells stably transfectedwith the long form of human leptin receptor. It also inhibitsvarious leptin effects in several in vitro bioassays.
- Proteinová synonyma:LEP
- Název proteinu/peptidu:Leptin
- Čistota:95% pure by SDS-PAGE
- Rozpustnost:It is recommended to reconstitute the lyophilized Leptinmutant in sterile 0.4% NaHCO3 adjusted to pH 8-9, notless than 100 g/ml, which can then be further diluted toother aqueous solutions. At low concentrations additionof a carrier protein (0.1% HSA or B
- Molekulární hmotnost:16 kDa
- Formulace:Lyophilized. Leptin is lyophilized from a solution containing 0.003 mM NaHCO3.