Nitrile gloves, uvex u-fit lite
Dodavatel: UVEX
Certifikáty
O tomto produktu
These disposable safety gloves provide reliable spray protection when handling chemicals such as acids, alkalis, solids or aqueous saline solutions.
- Good grip thanks to roughened fingertips
- Good mechanical resistance
- Silicone-free according to imprint test, accelerator-free
- Outstanding tactile feel
- Very good fit, extremely high flexibility
- Highly resistant to grease and oil
Typical applications include: Precision assembly work, inspection, short periods handling chemicals, paint shop (as splash protection), food processing, and gentle cleaning.
Certifications: EN ISO 374-1 Type C (K); AQL 1,5
Food contact approved
Food contact approved
Specifikace
- Catalog No:
- UVEX6059709
- UVEX6059707
- UVEX6059708
- Model:
- lite
- lite
- lite
- Obouruké/jednoruké:
- Obouruké
- Obouruké
- Obouruké
- Jednorázové/znovupoužitelné:
- Jednorázové
- Jednorázové
- Jednorázové
- Latex/bez latexu:
- Latex-free
- Latex-free
- Latex-free
- Provedení:
- Textured fingertips
- Textured fingertips
- Textured fingertips
- Norma EN:
- EN ISO 374-1 Type C (K)
- EN ISO 374-1 Type C (K)
- EN ISO 374-1 Type C (K)
- Schváleno pro styk s potravinami:
- +
- +
- +