Sterile containers, 250 ml, Corning®
Supplier: Corning
Certificates
About this item
PP Container, transparent, with snap-on HDPE lid.
- Graduated in both millilitres and ounces
- Certified non pyrogenic
- sterile
Serves as a beaker or storage container.
Caution: *Výrobky na této stránce nespadají do oblasti působnosti směrnice EU 2019/904.
Specifications
- Catalog No:
- 734-1817
- 525-3408
- 734-1818
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Certifikát ochrany životního prostředí:
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
Specifications
Related Information
Frequently Bought Together