Orders LinkContactUsLinkComponent
Reading magnifier, combi-plus
Reading magnifier, combi-plus
Aspherical reading magnifier, combi-plus, 2,8×, 100×75 mm
Katalogové číslo 630-1045
Dodavatel:   ESCHENBACH OPTIK
Reading magnifier, combi-plus
Katalogové číslo 630-1045
Dodavatel:  ESCHENBACH OPTIK
Reading magnifier, combi-plus
Aspherical reading magnifier, combi-plus, 2,8×, 100×75 mm
Přejít na:
Price
4 000,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Aspherical PXM® light-weight lens.
  • Ceratec® anti-scratch coating
  • Practical instrument
  • For individual use
This practical instrument can be used for reading or writing, either as a hand-held magnifier or as angled stand magnifier. It provides comfortable angled viewing, thanks to its fold-out metal feet.
Informace o dodávce: Supplied with black leather protective case with soft inner lining.
Specifikace
  • Zvětšení:
    2,8×
  • Popis:
    Aspherical reading magnifier, combi-plus
  • Š×H:
    100×75 mm