DNA methylation kit, express (shallow well)
Katalogové číslo ENZOENZ450030002
Dodavatel: Enzo Life Sciences
Číslo CAS:
Specifikace
- Bal.:1 KIT
- Popis:Express DNA methylation kit (shallow well), 2×96 wells
- Sekvence:02
Specifikace
O tomto produktu
The Express DNA Methylation Kit (shallow well) features high-throughput (96-well) bisulfite treatment and conversion of DNA for methylation analysis.
- Ready to use conversion reagent is added directly to DNA
- Easy to follow instructions
- High yield
DNA denaturation and bisulfite conversion processes are combined with added heat to facilitate rapid denaturation. Desulphonation and clean-up of the converted DNA is performed using a unique 96-well spin-plate. High yield, converted DNA is ideal for PCR, array, bisulfite and next generation sequencing.
Delivery: Kit contains Express conversion reagent, Methylation binding buffer, Methylation wash buffer concentrate, L-Desulphonation reagent, Methylation elution buffer, Express binding plate (shallow), Express conversion plates, collection plates and elution plates.